Phenazopyridine Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-138
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:136-40-3
- Description:Phenazopyridine Hydrochloride
- Size:10 mg
- Formula:C₁₁H₁₂ClN₅
- Molecular Weight:249.70 g/mol
- Cat. No.:77697-138