$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Mycophenolic Acid, LoGiCal, LGC Standards
Mycophenolic Acid, LoGiCal, LGC Standards
77697-133
:
LGC Standards
New Product
:
Mycophenolic Acid, LoGiCal, LGC Standards
77697-133
:
LGC Standards
:
LGCFOR1001.06
New Product
:
CAS No.:
24280-93-1
Description:
Mycophenolic Acid
Size:
10 mg
Formula:
C₁₇H₂₀O₆
Molecular Weight:
320.34 g/mol
Cat. No.:
77697-133