$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Norfenefrine Hydrochloride, LoGiCal, LGC Standards
Norfenefrine Hydrochloride, LoGiCal, LGC Standards
77697-132
:
LGC Standards
New Product
:
Norfenefrine Hydrochloride, LoGiCal, LGC Standards
77697-132
:
LGC Standards
:
LGCFOR0973.00
New Product
:
CAS No.:
4779-94-6
Description:
Norfenefrine Hydrochloride
Size:
10 mg
Formula:
C₈H₁₁NO₂
Molecular Weight:
189.64 g/mol
Cat. No.:
77697-132