$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Orphenadrine Hydrochloride, LoGiCal, LGC Standards
Orphenadrine Hydrochloride, LoGiCal, LGC Standards
77697-123
:
LGC Standards
New Product
:
Orphenadrine Hydrochloride, LoGiCal, LGC Standards
77697-123
:
LGC Standards
:
LGCFOR0801.00
New Product
:
CAS No.:
341-69-5
Description:
Orphenadrine Hydrochloride
Size:
10 mg
Formula:
C₁₈H₂₃NO
Molecular Weight:
305.85 g/mol
Cat. No.:
77697-123