Methoxyphenamine Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-115
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:5588-10-3
- Description:Methoxyphenamine Hydrochloride
- Size:10 mg
- Formula:C₁₁H₁₈ClNO
- Molecular Weight:215.721 g/mol
- Cat. No.:77697-115