Salmeterol Xinafoate, LoGiCal, LGC Standards
Catalog # 77697-105
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:94749-08-3
- Description:Salmeterol Xinafoate
- Size:10 mg
- Formula:C₂₅H₃₇NO₄·C₁₁H₈O₃
- Molecular Weight:603.76 g/mol
- Cat. No.:77697-105