Finasteride, LoGiCal, LGC Standards
Catalog # 77697-096
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:98319-26-7
- Description:Finasteride
- Size:10 mg
- Formula:C₂₃H₃₆N₂O₂
- Molecular Weight:372.55 g/mol
- Cat. No.:77697-096