Oxymetazoline Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-067
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2315-02-8
- Description:Oxymetazoline Hydrochloride
- Size:10 mg
- Formula:C₁₆H₂₄N₂O·HCl
- Molecular Weight:296.839 g/mol
- Cat. No.:77697-067