Mianserin Hydrochloride, LoGiCal, LGC Standards
Catalog # 77697-065
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:21535-47-7
- Description:Mianserin Hydrochloride
- Size:10 mg
- Formula:C₁₈H₂₀N₂
- Molecular Weight:300.83 g/mol
- Cat. No.:77697-065