Fluphenazine Dihydrochloride, LoGiCal, LGC Standards
Catalog # 77697-063
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:146-56-5
- Description:Fluphenazine Dihydrochloride
- Size:10 mg
- Formula:C₂₂H₂₈Cl₂F₃N₃OS
- Molecular Weight:510.441 g/mol
- Cat. No.:77697-063