$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Nortetrazepam, LoGiCal, LGC Standards
Nortetrazepam, LoGiCal, LGC Standards
77697-056
:
LGC Standards
New Product
:
Nortetrazepam, LoGiCal, LGC Standards
77697-056
:
LGC Standards
:
LGCFOR0091.03
New Product
:
CAS No.:
10379-11-0
Description:
Nortetrazepam
Size:
10 mg
Cat. No.:
77697-056