Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
N-Desmethyldiltiazem Hydrochloride, LoGiCal, LGC Standards
N-Desmethyldiltiazem Hydrochloride, LoGiCal, LGC Standards
N-Desmethyldiltiazem Hydrochloride, LoGiCal, LGC Standards
77697-050
:
LGC Standards
New Product
:
N-Desmethyldiltiazem Hydrochloride, LoGiCal, LGC Standards
77697-050
:
LGC Standards
:
LGCFOR0068.03
New Product
:
CAS No.:
130606-60-9
Description:
N-Desmethyldiltiazem Hydrochloride
Size:
10 mg
Cat. No.:
77697-050