Diphenyl(piperidin-4-yl)methanol (Azacyclonol), LoGiCal, LGC Standards
Catalog # 77697-047
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:132-20-7
- Description:Diphenyl(piperidin-4-yl)methanol (Azacyclonol)
- Size:10 mg
- Formula:C₂₀H₂₄N₂O₄
- Molecular Weight:356.422 g/mol
- Cat. No.:77697-047