Etizolam 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77697-018
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:40054-69-1
- Description:Etizolam 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₇H₁₅ClN₄S
- Molecular Weight:342.845 g/mol
- Cat. No.:77697-018