Pentobarbital 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77697-000
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:76-74-4
- Description:Pentobarbital 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₁H₁₈N₂O₃
- Molecular Weight:226.28 g/mol
- Cat. No.:77697-000