$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Methaqualone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Methaqualone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77696-998
:
LGC Standards
New Product
:
Methaqualone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77696-998
:
LGC Standards
:
LGCAMP1356.00-02
New Product
:
CAS No.:
72-44-6
Description:
Methaqualone 0.1 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77696-998