Hexobarbital 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-994
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:56-29-1
- Description:Hexobarbital 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₂H₁₆N₂O₃
- Molecular Weight:236.27 g/mol
- Cat. No.:77696-994