LSD (Lysergic Acid Diethylamide) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-992
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:50-37-3
- Description:LSD (Lysergic Acid Diethylamide) 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₀H₂₅N₃O
- Cat. No.:77696-992