para-Fluorophenylpiperazine Dihydrochloride (1-(4-Fluorophenyl)piperazine Dihydrochloride) 1.0 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal, LGC Standards
77696-987
New Product
:
- CAS No.:64090-19-3
- Description:para-Fluorophenylpiperazine Dihydrochloride (1-(4-Fluorophenyl)piperazine Dihydrochloride) 1.0 mg/ml in Dimethyl Sulfoxide (as free base)
- Size:1 ml
- Formula:C₁₀H₁₅Cl₂FN₂
- Molecular Weight:253.15 g/mol
- Cat. No.:77696-987