Order Entry
Puerto Rico
ContactUsLinkComponent
JWH-210 ((4-Ethylnaphthalen-1-yl)(1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
JWH-210 ((4-Ethylnaphthalen-1-yl)(1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-978
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
JWH-210 ((4-Ethylnaphthalen-1-yl)(1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-978
Supplier:  LGC Standards
Supplier Number:  LGCAMP1275.67-11
New Product
CAS Number:  

Specifications

  • CAS No.:
    824959-81-1
  • Description:
    JWH-210 ((4-Ethylnaphthalen-1-yl)(1-pentylindol-3-yl)methanone) 1.0 mg/ml in Acetonitrile
  • Size:
    1 ml
  • Cat. No.:
    77696-978

Specifications

About this item