Methedrone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77696-969
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:879665-92-6
- Description:Methedrone Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₁H₁₆ClNO₂
- Molecular Weight:229.70 g/mol
- Cat. No.:77696-969