Haloperidol 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-962
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52-86-8
- Description:Haloperidol 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₂₁H₂₃ClFNO₂
- Molecular Weight:375.87 g/mol
- Cat. No.:77696-962