Quetiapine Fumarate 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77696-961
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:111974-72-2
- Description:Quetiapine Fumarate 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₁H₂₅N₃O₂S
- Molecular Weight:441.55 g/mol
- Cat. No.:77696-961