$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Phentermine 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Phentermine 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-944
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Phentermine 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-944
Supplier:
LGC Standards
Supplier Number:
LGCAMP0962.02-01
New Product
CAS Number:
Specifications
CAS No.:
122-09-8
Description:
Phentermine 1.0 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77696-944
Specifications
About this item
Product Details
Product Details