Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pholcodine-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Pholcodine-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Pholcodine-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-927
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Pholcodine-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-927
Supplier:
LGC Standards
Supplier Number:
LGCAMP0733.80-01
New Product
CAS Number:
Specifications
Description:
Pholcodine-D3 1.0 mg/ml in Methanol
Size:
1 ml
Cat. No.:
77696-927
Specifications
About this item
Product Details
Product Details