Donepezil Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77696-926
New Product
:
- CAS No.:120011-70-3
- Description:Donepezil Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₄H₂₉NO₃
- Molecular Weight:415.96 g/mol
- Cat. No.:77696-926