Etofylline 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-920
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:519-37-9
- Description:Etofylline 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₉H₁₂N₄O₃
- Molecular Weight:224.216 g/mol
- Cat. No.:77696-920