Indometacin 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-917
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:53-86-1
- Description:Indometacin 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₁₉H₁₆ClNO₄
- Molecular Weight:357.80 g/mol
- Cat. No.:77696-917