Methyltestosterone (17alpha-Methyltestosterone) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-916
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:58-18-4
- Description:Methyltestosterone (17alpha-Methyltestosterone) 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₀H₃₀O₂
- Molecular Weight:302.46 g/mol
- Cat. No.:77696-916