$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
rac-Methadone-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
rac-Methadone-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-911
:
LGC Standards
New Product
:
rac-Methadone-D3 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-911
:
LGC Standards
:
LGCAMP0531.80-01
New Product
:
CAS No.:
60263-63-0
Description:
rac-Methadone-D3 1.0 mg/ml in Methanol
Size:
1 ml
Formula:
C₂₁H₂₄D₃NO
Cat. No.:
77696-911