$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
rac-Methadone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
rac-Methadone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77696-910
:
LGC Standards
New Product
:
rac-Methadone 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
77696-910
:
LGC Standards
:
LGCAMP0531.08-02
New Product
:
CAS No.:
76-99-3
Description:
rac-Methadone 0.1 mg/ml in Methanol
Size:
1 ml
Formula:
C₂₁H₂₇NO
Cat. No.:
77696-910