rac-Methadone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-909
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:76-99-3
- Description:rac-Methadone 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₂₁H₂₇NO
- Cat. No.:77696-909