Isosorbide Mononitrate 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal, LGC Standards
Catalog # 77696-903
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:16051-77-7
- Description:Isosorbide Mononitrate 1.0 mg/ml in Dimethyl Sulfoxide
- Size:1 ml
- Formula:C₆H₉NO₆
- Molecular Weight:191.139 g/mol
- Cat. No.:77696-903