Mesterolone 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-902
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1424-00-6
- Description:Mesterolone 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₀H₃₂O₂
- Molecular Weight:304.47 g/mol
- Cat. No.:77696-902