$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Risperidone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Risperidone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-901
:
LGC Standards
New Product
:
Risperidone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-901
:
LGC Standards
:
LGCAMP0491.00-01
New Product
:
CAS No.:
106266-06-2
Description:
Risperidone 1.0 mg/ml in Methanol
Size:
1 ml
Formula:
C₂₃H₂₇FN₄O₂
Molecular Weight:
410.49 g/mol
Cat. No.:
77696-901