Indapamide 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-898
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:26807-65-8
- Description:Indapamide 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₆H₁₆ClN₃O₃S
- Molecular Weight:365.83 g/mol
- Cat. No.:77696-898