Promethazine Sulphoxide 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-895
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:7640-51-9
- Description:Promethazine Sulphoxide 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₇H₂₀N₂OS
- Molecular Weight:300.42 g/mol
- Cat. No.:77696-895