O-Desmethylvenlafaxine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-893
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:93413-62-8
- Description:O-Desmethylvenlafaxine 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₆H₂₅NO₂
- Molecular Weight:263.38 g/mol
- Cat. No.:77696-893