Melperone 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-889
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3575-80-2
- Description:Melperone 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₆H₂₂FNO
- Molecular Weight:263.36 g/mol
- Cat. No.:77696-889