$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Pethidine (Meperidine) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Pethidine (Meperidine) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-884
:
LGC Standards
New Product
:
Pethidine (Meperidine) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-884
:
LGC Standards
:
LGCAMP0315.11-01
New Product
:
CAS No.:
57-42-1
Description:
Pethidine (Meperidine) 1.0 mg/ml in Methanol
Size:
1 ml
Formula:
C₁₅H₂₁NO₂
Cat. No.:
77696-884