Molsidomine 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-879
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:25717-80-0
- Description:Molsidomine 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₉H₁₄N₄O₄
- Molecular Weight:242.24 g/mol
- Cat. No.:77696-879