Phenobarbital 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-876
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:50-06-6
- Description:Phenobarbital 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₂H₁₂N₂O₃
- Molecular Weight:232.24 g/mol
- Cat. No.:77696-876