Spironolactone 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-868
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52-01-7
- Description:Spironolactone 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₄H₃₂O₄S
- Molecular Weight:416.58 g/mol
- Cat. No.:77696-868