Flunarizine Dihydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
77696-867
New Product
:
- CAS No.:30484-77-6
- Description:Flunarizine Dihydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₂₆H₂₆F₂N₂·2HCl
- Molecular Weight:477.40 g/mol
- Cat. No.:77696-867