$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Naproxen 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Naproxen 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-865
:
LGC Standards
New Product
:
Naproxen 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
77696-865
:
LGC Standards
:
LGCAMP0152.00-01
New Product
:
CAS No.:
22204-53-1
Description:
Naproxen 1.0 mg/ml in Methanol
Size:
1 ml
Formula:
C₁₄H₁₄O₃
Molecular Weight:
230.26 g/mol
Cat. No.:
77696-865