Oxprenolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77696-861
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:6452-73-9
- Description:Oxprenolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₅H₂₄ClNO₃
- Molecular Weight:301.82 g/mol
- Cat. No.:77696-861