Diphenyl(piperidin-4-yl)methanol (Azacyclonol; Pipradrol 4-Isomer) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-843
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:132-20-7
- Description:Diphenyl(piperidin-4-yl)methanol (Azacyclonol;Pipradrol 4-Isomer) 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₂₀H₂₄N₂O₄
- Molecular Weight:356.422 g/mol
- Cat. No.:77696-843