Metamizole Sodium Monohydrate 1.0 mg/ml in Dimethyl Sulfoxide (as anhydrous free acid), LoGiCal, LGC Standards
Catalog # 77696-842
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:5907-38-0
- Description:Metamizole Sodium Monohydrate 1.0 mg/ml in Dimethyl Sulfoxide (as anhydrous free acid)
- Size:1 ml
- Formula:C₁₃H₁₆N₃NaO₄S·H₂O
- Molecular Weight:351.36 g/mol
- Cat. No.:77696-842