Meclofenoxate Hydrochloride 1.0 mg/ml in Acetonitrile (as free base), LoGiCal, LGC Standards
Catalog # 77696-840
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3685-84-5
- Description:Meclofenoxate Hydrochloride 1.0 mg/ml in Acetonitrile (as free base)
- Size:1 ml
- Formula:C₁₂H₁₆ClNO₃
- Molecular Weight:294.17 g/mol
- Cat. No.:77696-840