Propranolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal, LGC Standards
Catalog # 77696-839
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:318-98-9
- Description:Propranolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
- Size:1 ml
- Formula:C₁₆H₂₁NO₂·HCl
- Molecular Weight:295.81 g/mol
- Cat. No.:77696-839